Learn More
Sulfuric Acid, 0.5M, Ph. Eur., Reagecon™
BIOPHARMA
Product Code. 11434819 Shop All Reagecon Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
1L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11434819 1 L 1L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11434819 Supplier Reagecon Supplier No. 3007800
Specifications
Concentration 0.5M
Packaging 1L
Quantity 1 L
compliance-icons
Product Identifier
  • SULPHURIC ACID 0.5M
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: sulphuric acid