missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sulindac

80.60€

Chemical Identifiers

CAS 38194-50-2
Formula Weight 356.41g/mol
Synonym cis-5-Fluoro-2-methyl-1-[4-(methylsulfinyl)benzylidene]indene-3-acetic acid; MK-231
Content And Storage -30°C to -10°C
Format Powder
For Use With (Application) A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase-1 and induces apoptosis in HT-29 cells
Description Yellow
Melting Point 182°C to 185°C
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15404299
SDS View Documents
Thermo Scientific™
J61772.06
5 g
80.60€
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase-1 and induces apoptosis in HT-29 cells.
Specifications

Chemical Identifiers

38194-50-2
cis-5-Fluoro-2-methyl-1-[4-(methylsulfinyl)benzylidene]indene-3-acetic acid; MK-231
Powder
Yellow
356.41g/mol
-30°C to -10°C
A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase-1 and induces apoptosis in HT-29 cells
182°C to 185°C

Specifications

Yellow
Powder
38194-50-2
356.41g/mol
182°C to 185°C
-30°C to -10°C
A non-steroidal anti-inflammatory drug (NSAID) that inhibits cyclooxygenase-1 and induces apoptosis in HT-29 cells
C20H17FO3S
cis-5-Fluoro-2-methyl-1-[4-(methylsulfinyl)benzylidene]indene-3-acetic acid; MK-231
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 253-819-2

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.