Learn More
Sulfur Trioxide Triethylamine Complex, TRC
Specifications
Specifications
| Chemical Name or Material | Sulfur Trioxide Triethylamine Complex |
| CAS | 761-01-3 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C6H15NO3S |
| Quantity | 10 g |
| Synonym | Triethylamine compounded with SO3, Triethylamine compounded with sulfur trioxide, N,N-Diethyl-ethanamine compounded with sulfur trioxide, Sulfur Trioxide compounded with triethylamine, NSC 68185, NSC 8168, Sulfur trioxide-triethylamine compound, Sulfur trioxide-triethylamine complex, Triethylamine-sulfur trioxide compound, Triethylamine-sulfur trioxide complex |
| InChI Formula | InChI=1S/C6H15N.O3S/c1-4-7(5-2)6-3;1-4(2)3/h4-6H2,1-3H3; |
| SMILES | CCN(CC)CC.O=S(=O)=O |
| IUPAC Name | N,N-diethylethanamine;sulfur trioxide |
| Molecular Weight (g/mol) | 181.25 |
| Show More |