Learn More
Sulfosuccinic Acid Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | Sulfosuccinic Acid Sodium Salt |
| CAS | 29454-16-8 |
| Molecular Formula | C4H5NaO7S |
| Quantity | 5 g |
| Synonym | 2-Sulfo-butanedioic Acid Sodium Salt, Sulfo-succinic Acid Monosodium Salt |
| InChI Formula | InChI=1S/C4H6O7S.Na/c5-3(6)1-2(4(7)8)12(9,10)11;/h2H,1H2,(H,5,6)(H,7,8)(H,9,10,11);/q;+1/p-1 |
| SMILES | C(C(C(=O)O)S(=O)(=O)[O-])C(=O)O.[Na+] |
| IUPAC Name | sodium 1,2-dicarboxyethane-1-sulfonate |
| Molecular Weight (g/mol) | 220.13 |
| Formula Weight | 219.97 |