Learn More
Sulfinyldibenzene, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 945-51-7 |
| Molecular Formula | C12H10OS |
| MDL Number | MFCD00002085 |
| Quantity | 100 g |
| SMILES | O=S(C1=CC=CC=C1)C2=CC=CC=C2 |
| Molecular Weight (g/mol) | 202.27 |
| Percent Purity | 98% |
| Chemical Name or Material | Sulfinyldibenzene |