Learn More
Sulfanilamide, Extra Pure, SLR, Fisher Chemical
Product Code. 10733811 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10733811 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10733811 Supplier Fisher Chemical Supplier No. S/8660/50
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 63-74-1 | C6H8N2O2S | 172.202 g/mol
250g, Plastic powder jar
TRUSTED_SUSTAINABILITY
Specifications
Physical Form Solid
CAS 63-74-1
pH 5.8 to 6.1
Melting Point 163°C
Packaging Plastic Powder Jar
Color White
Quantity 250 g
Molecular Formula C6H8N2O2S
MDL Number 7939
InChI Key FDDDEECHVMSUSB-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1N)S(=O)(=O)N
IUPAC Name 4-aminobenzenesulfonamide
Molecular Weight (g/mol) 172.202
PubChem CID 5333
ChEBI CHEBI:45373
Formula Weight 172.2g/mol
Show More Show Less
compliance-icons
Product Identifier
  • Sulfanilamide
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.