Learn More
Sulfamerazine-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Sulfamerazine-d4 |
| CAS | 1020719-84-9 |
| Molecular Formula | C11 2H4 H8 N4 O2 S |
| Quantity | 1 mg |
| Synonym | Benzene-2,3,5,6-d4-sulfonamide','4-amino-N-(4-methyl-2-pyrimidinyl)-, Sulfamerazine D4 |
| InChI Formula | InChI=1S/C11H12N4O2S/c1-8-6-7-13-11(14-8)15-18(16,17)10-4-2-9(12)3-5-10/h2-7H,12H2,1H3,(H,13,14,15)/i2D,3D,4D,5D |
| SMILES | [2H]c1c([2H])c(c([2H])c([2H])c1N)S(=O)(=O)Nc2nccc(C)n2 |
| IUPAC Name | 4-amino-2,3,5,6-tetradeuterio-N-(4-methylpyrimidin-2-yl)benzenesulfonamide |
| Molecular Weight (g/mol) | 268.33 |
| Formula Weight | 268.0932 |