missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Picolinafen-d4 (4-fluorophenyl-d4), CDN
Click to view available options
Quantity:
0.01 g
0.05 g
Unit Size:
0.01g
0.05g
Specifications
Specifications
| Chemical Name or Material | Picolinafen-d4 |
| Molecular Formula | C19D4H8F4N2O2 |
| Quantity | 0.01 g |
| Synonym | AC 900001-d4, N-(4-Fluorophenyl)-2-[(3-trifluoromethylphenyl)oxy]pyridine-6-carboxamide-d4, N-(4-Fluorophenyl)-6-[3-(trifluoromethyl)phenoxy]-2-pyridinecarboxamide-d4, Sniper-d4 |
| InChI Formula | InChI=1 S/C19H12F4N2O2/c20-13-7-9-14(10-8-13)24-18(26)16-5-2-6-17(25-16)27-15-4-1-3-12(11-15)19(21,22)23/h1-11 H,(H,24,26)/i7D,8 D,9 D,10 D |
| SMILES | [2 H]c1c([2 H])c(NC(=O)c2cccc(Oc3cccc(c3)C(F)(F)F)n2)c([2 H])c([2 H])c1F |
| IUPAC Name | N-(2,3,5,6-tetradeuterio-4-fluoro-phenyl)-6-[3-(trifluoromethyl)phenoxy]pyridine-2-carboxamide |
| Molecular Weight (g/mol) | 380.33 |
| Formula Weight | 380.109 g/mol |
| Percent Purity | 99 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction