Learn More
Sulfaguanidine, TRC
Product Code. 30134700
Change view
Click to view available options
Quantity:
50 mg
1 g
10 g
Unit Size:
10g
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134700 50 mg 50mg
30137603 10 g 10g
17914705 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134700 Supplier Toronto Research Chemicals Supplier No. TRCS68898550MG
 Restricted Item
Estimated Shipment: 04-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 57-67-0
Molecular Formula C7 H10 N4 O2 S
Molecular Weight (g/mol) 214.24
Synonym (4-Aminophenylsulfonyl)guanidine, Sulfaguanidine, Sulfadimidine Imp. C (Pharmeuropa), N1-(Diaminomethylene)sulfanilamide, 1-Sulfanilylguanidine, 1-[(p-Aminophenyl)sulfonyl]guanidine, 4-Amino-N-(aminoiminomethyl)benzenesulfonamide, 4-Aminobenzenesulfonylguanidine, 4-Aminophenylsulfonylguanidine, A 307, Abiguanil, Aterian, Diacta, Emerin (pharmaceutical), Ganidan, Guamide, Guanicil, Guanidan, N-Guanylsulfanilamide, N1-Amidinosulfanilamide, N1-Guanidylsulfanilamide, N1-Guanylsulfanilamide, NSC 14041, Orgaguanidon, RP 2275, Resulfon, Ruocid, Shigatox, Suganyl, Sulfaguanidin, Sulfaguanil, Sulfaguine, Sulfanilguanidine, Sulfanilylguanidine, Sulfentidine, Sulfoguanidine, Sulfoguanil, Sulfoguanyl, Sulfoguenil, Sulgin, Sulphaguanidine, [(p-Aminophenyl)sulfonyl]guanidine, p-Aminobenzenesulfoguanidide, p-Aminobenzenesulfonylguanidine
IUPAC Name 1-(4-aminophenyl)sulfonylguanidine
SMILES NC(=N)NS(=O)(=O)c1ccc(N)cc1
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C7H10N4O2S/c8-5-1-3-6(4-2-5)14(12,13)11-7(9)10/h1-4H,8H2,(H4,9,10,11)
Formula Weight 214.0524
Percent Purity >95%
Chemical Name or Material Sulfaguanidine
Grade HPLC