Learn More
Sulfadiazine-d4, TRC
Specifications
Specifications
| Chemical Name or Material | Sulfadiazine D4 |
| CAS | 1020719-78-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10 D4 H6 N4 O2 S |
| Quantity | 10 mg |
| Synonym | 4-Amino-N-2-pyrimidinylbenzene-2,3,5,6-d4-sulfonamide, Benzene-2,3,5,6-d4-sulfonamide','4-amino-N-2-pyrimidinyl-, Sulfadiazine D4 |
| InChI Formula | InChI=1S/C10H10N4O2S/c11-8-2-4-9(5-3-8)17(15,16)14-10-12-6-1-7-13-10/h1-7H,11H2,(H,12,13,14)/i2D,3D,4D,5D |
| SMILES | [2H]c1c([2H])c(c([2H])c([2H])c1N)S(=O)(=O)Nc2ncccn2 |
| IUPAC Name | 4-amino-2,3,5,6-tetradeuterio-N-pyrimidin-2-ylbenzenesulfonamide |
| Molecular Weight (g/mol) | 254.302 |
| Show More |