missing translation for 'onlineSavingsMsg'
Learn More

Sulfachloropyridazine sodium, 97%, Thermo Scientific™

Product Code. p-7077711 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15502333 1 g 1g
15512333 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15502333 Supplier Thermo Scientific Acros Supplier No. 458060010
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 23282-55-5
Molecular Formula C10H8ClN4NaO2S
Molecular Weight (g/mol) 306.7
InChI Key GHQJXOBLWKFNAD-UHFFFAOYSA-N
PubChem CID 87077948
IUPAC Name 4-amino-N-(6-chloropyridazin-3-yl)benzenesulfonamide;sodium
SMILES C1=CC(=CC=C1N)S(=O)(=O)NC2=NN=C(C=C2)Cl.[Na]

Specifications

Melting Point 262.0°C to 270.0°C
Loss on Drying 2% max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 306.7
Percent Purity 97%
Chemical Name or Material Sulfachloropyridazine sodium
compliance-icons
Product Identifier
  • Sulphachloropyridazine sodium
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Sodium N-(6-chloropyridazin-3-yl)sulphanilamidate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.