Learn More
R&D Systems™ Substrate Reagent Pack
Click to view available options
Quantity:
1 Ea.
Unit Size:
Pack of 16
Specifications
Specifications
| Quantity | 1 Ea. |
| For Use With (Application) | ELISA |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |