Learn More
BUCHI Glass Oven Sublimation Accessories
Product Code. 10027341 Shop All Buchi Products
Change view
Click to view available options
For Use With (Equipment):
B 580
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
10027341 B 580 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10027341 Supplier BUCHI Supplier No. 037133
Sublimation accessory for BUCHI B-580
Sublimation finger Glass tube
  • Includes bent hose barb and cap nut GL14
  • For sublimation accessory (037133)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) B 580
Type Sublimation Accessory
Includes Flange Ring, O-Ring, Stopcock
Material Glass