Learn More
Memmert™ Subframe for UF55plus, Adjustable up to 600 mm
Product Code. 17579475 Shop All Memmert Products
Click to view available options
Description:
Adjustable
Unit Size:
Each
Product Code. 17579475
Brand: Memmert™ UF55plusB29747
Only null left
Add to basket
Add to basket
Adjustable subframe for Memmert Ovens.
  • Adjustable in height
Specifications
Description Adjustable
For Use With (Equipment) UF55plus Oven
Type Subframe
Height (Metric) Up to 600 mm