Learn More
Memmert Subframe height 130mm (only with corresponding stacking set) for HCP105
Brand:  Memmert HCP105B33508
Subframe height 130mm (only with corresponding stacking set) for HCP105
Product Code. 16334787
  • ONLINE EXCLUSIVE PRICE
    391.00€
    355.00€ / Each
    Save 36.00€
    9% Off
Only null left
Estimated Shipment: 11-03-2026