Learn More
Memmert Subframe adjustable in height up to 622mm, for HCP105
Brand:  Memmert HCP105B33505
Subframe adjustable in height up to 622mm, for HCP105
Product Code. 16324787
  • ONLINE EXCLUSIVE PRICE
    415.00€
    377.00€ / Each
    Save 38.00€
    9% Off
Only null left
Estimated Shipment: 12-03-2026