Learn More
Hoefer SUB Evo Horizontal Electrophoresis System
Product Code. 32180971
Change view
Click to view available options
Capacity:
216 Samples
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Unit Size
32180971 216 Samples Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 32180971 Supplier Hoefer Supplier No. SUB Evo
  • ONLINE EXCLUSIVE PRICE
    869.79€
    741.66€ / Pack of 1
    Save 128.13€
    14% Off
Estimated Shipment: 21-10-2026
Supports high-throughput nucleic acid analysis with capacity for up to two 96-well plates
  • High throughput: Run up to 216 samples, compatible with 8- or 12-channel pipettes
  • Sequential 8-channel loading: Efficient operation and smooth result review
  • Flexible gel setup: Supports gels up to 25 x 17 cm with multiple trays and combs
  • Plug-free lid: Smooth operation while protecting electrodes
  • UV-transparent tray: Allows gel imaging without removal
Specifications
Certifications/Compliance CE
Capacity 216 Samples
Detection Method Not Applicable
For Use With (Application) Nucleic Acid Analysis
For Use With (Equipment) SUB Evo Horizontal Electrophoresis System
Gel Compatibility Not Applicable
Gel Size 16 x 17.5 cm
Max. Current 500 mA
Running Time 30 to 60 min.
Timer Not Applicable
Type Horizontal Agarose Electrophoresis Unit
Gel Dimensions 25 x 17 cm, 17.5 x 17 cm, 7.5 x 17 cm
No. of Wells 18+36 Wells
Quantity 1 Pc.
Thickness (Metric) Teeth 1.5 mm
Volume (Metric) Buffer 900 mL
Dimensions (L x W x H) 42.2 x 19.7 x 7.2 cm
Max. Voltage 1000 V
Voltage 100/240 V
Frequency 50/60 Hz