missing translation for 'onlineSavingsMsg'
Learn More

(+/-)-Styrene oxide, 98+%

Product Code. p-7044695
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312067 100 g 100g
11322067 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11312067 Supplier Thermo Scientific Alfa Aesar Supplier No. L07821.22
Add to basket
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 96-09-3 | C8H8O | 120.151 g/mol

Styrene oxide is used as an intermediate in medicine and perfume.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Styrene oxide is used as an intermediate in medicine and perfume.

Solubility
Soluble in water.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-09-3
Molecular Formula C8H8O
Molecular Weight (g/mol) 120.151
MDL Number MFCD00005121
InChI Key AWMVMTVKBNGEAK-UHFFFAOYSA-N
Synonym styrene oxide, phenyloxirane, 1,2-epoxyethylbenzene, phenylethylene oxide, epoxystyrene, styrene epoxide, styryl oxide, oxirane, phenyl, 1-phenyloxirane, phenethylene oxide
PubChem CID 7276
ChEBI CHEBI:17907
IUPAC Name 2-phenyloxirane
SMILES C1C(O1)C2=CC=CC=C2

Specifications

Melting Point -37°C
Density 1.053
Boiling Point 193°C to 196°C
Flash Point 74°C (165°F)
Refractive Index 1.535
Quantity 100 g
UN Number UN2810
Beilstein 108582
Solubility Information Soluble in water.
Formula Weight 120.15
Percent Purity ≥98%
Chemical Name or Material (+/-)-Styrene oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.