missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Styrene-alpha-d1, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Styrene-alpha-d1 |
| CAS | 1193-80-2 |
| Molecular Formula | C8 2 H H7 |
| Quantity | 1 g |
| Synonym | Benzene, ethenyl-1-d-, Styrene-α-d (6 CI,7 CI,8 CI), Ethenyl-1-dbenzene, Styrene-α-d1, α-Deuteriostyrene, α-Deuterostyrene, Styrene-α-D1 |
| InChI Formula | InChI=1 S/C8H8/c1-2-8-6-4-3-5-7-8/h2-7 H,1H2/i2D |
| SMILES | [2 H]C(=C)c1ccccc1 |
| IUPAC Name | 1-deuterioethenylbenzene |
| Molecular Weight (g/mol) | 105.16 |
| Formula Weight | 105.0689 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction