Learn More
Cole-Parmer™ Clip Plate for Rotator Drum
Product Code. 12063324
Click to view available options
For Use With (Equipment):
For 10 Tubes
For 12 Tubes
For 7 Tubes
For 8 Tubes
Unit Size:
Pack of 2
Product Code. 12063324
Brand: Cole-Parmer™ 5160094
Optimize performance of your DR-200 Series Rotating Shaker System with this Clip Plate for Rotator Drums.
  • Change your rotator's capacity with interchangeable drums
  • Clip plates hold various size tubes
Drums are compatible with Cole-Parmer™ DR-200 Series Rotating Shaker System Drives (sold separately). Use drive unit in conjunction with a wide range of drums and clip plates designed to take different sizes and types of vessels.
Specifications
Product Type Tube Rack
Holds Tubes (24 mm)
Positions 7
For Use With (Equipment) For 7 Tubes
Diameter (Metric) Opening 24 mm