Learn More
Cole-Parmer™ Stuart™ Pressmatic Polypropylene Dispensers
Product Code. 12060892
Change view
Click to view available options
Volume (Metric):
10 to 60 mL
2 to 10 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
12060892 2 to 10 mL Each
12040902 10 to 60 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12060892 Supplier Cole-Parmer™ Supplier No. 1011276
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Accuracy <0.7%
Product Type Bottletop Dispenser
Material Polypropylene
Volume (Metric) 2 to 10 mL
For Use With (Equipment) Bottle
Increments 0.25 mL