Learn More
Cole-Parmer™ Stuart™ Gabled Lid
Product Code. 12757969
Click to view available options
For Use With (Equipment):
12L Bath
18 and 26L Bath
48L Bath
8L Bath
Unit Size:
Each
Product Code. 12757969
Brand: Cole-Parmer™ 1212296
Enhance the performance of your Techne baths and circulators
  • Lids, trays, temperature probes, and other accessories for your bath.
  • Manufactured from stainless steel and available to fit 8 L baths to help prevent evaporation losses.
Specifications
For Use With (Equipment) 12L Bath