Learn More
Cole-Parmer™ Dry Block Heater Cover
Product Code. p-7182441
Click to view available options
Size:
2 Biock System
3 Block or Dual Control
Unit Size:
Each
Product Code. 11436598
Brand: Cole-Parmer™ 3661096
Only null left
Add to basket
Add to basket
Optimize performance of your block heaters with this cover.
  • The heater covers ensure temperatures are maintained in your dry block heaters
  • Model 36610-96 cover is suitable for use with two block heaters
  • Model 36610-97 is suitable for use with three block or dual control heaters
  • The extraction tool, 36610-98, makes removing blocks from the heater quick and effortless
Specifications
For Use With (Equipment) 2-block System
Type Safety Cover
Size 2 Biock System