Learn More
Cole-Parmer™ Stuart™ Block Heater Insert
Product Code. p-8012840
Click to view available options
Diameter (Metric) Well:
7.8 mm
10 mm
10.5 mm
10.7 mm
12.6 mm
13.5 mm
16.5 mm
19.5 mm
25.5 mm
28 mm
30 mm
61 mm
Depth (Metric) Well:
14 mm
33 mm
47 mm
No. of Places:
4
6
8
12
20
30
48
Unit Size:
Each
Product Code. 11496588
Brand: Cole-Parmer™ 3661059
Use these blocks for a variety of applications. Stuart™ Anodized Aluminum Blocks may be used in any combination.
  • For use with Stuart block heaters
  • Manufactured from anodized aluminum
  • Separate hole accommodates a thermometer if desired
Specifications
Description Holds 4 x 30 mm tubes
Holds 4 x 30 mm tubes
Material Anodized Aluminum
No. of Places 4
Type Block
Depth (Metric) Well 47 mm
Diameter (English) Well 1.18 in.
Diameter (Metric) Well 30 mm