Learn More
Cole-Parmer™ Stuart™ Analogue Tube Roller
Product Code. 11416498
Click to view available options
Includes:
6 Rollers
9 Rollers
Depth (Metric):
255 mm
375 mm
Unit Size:
Each
Product Code. 11416498
Brand: Cole-Parmer™ 0465304
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Provides a gentle, but highly efficient, rocking and rolling action. Stuart™ Analogue Tube Roller Units can be used in incubators up to 60°C and humidity up to 80%, or in cold rooms down to 4°C.
Provides a gentle, but highly efficient, rocking and rolling action. The six rollers design has a small space-saving footprint, while the nine roller unit has a higher capacity.
  • Control is through a simple on/off switch and the unit will run continuously at the fixed speed of 33rpm
  • Recommended for mixing blood samples, viscous substances and liquid-solid suspensions where minimum aeration is required.
  • Units can be used in incubators up to 60°C and humidity up to 80%, or in cold rooms down to 4°C. Both roller mixers are robustly constructed and designed for easy cleaning, having plastic rollers and a drip tray to collect accidental spillages.
  • Most sizes of tubes, Bijoux, Universals and bottles can be accommodated, up to 100mm diameter.
Specifications
Speed Range 33 rpm
Electrical Requirements 230 V, 50 Hz, 50 W
Voltage 230 V
Wattage 50 W
Frequency 50 Hz
Depth (Metric) 375 mm
Height (Metric) 110 mm
Length (Metric) 340 mm
Width (Metric) 595 mm
Controller Type Analogue
Model SRT9
For Use With (Application) Mixing blood samples, viscous substances and liquid-solid suspensions where minimum aeration is required
Humidity Range Up to 80%
Includes 9 Rollers
Packaging Quantity 1 Ea.