Learn More
Techne™ Dri-Block™ Aluminum Heating Block Insert
Click to view available options
No. of Places:
2
Unit Size:
Each
Description
- Versatile range of interchangeable heating blocks work with many size sample tubes and plates
- Heaters sold separately
Specifications
Specifications
| Description | Conical wells |
| Material | Anodized Aluminum |
| No. of Places | 2 |
| Type | Insert Block |
| Depth (English) Well | 1.50 in. |
| Depth (Metric) Well | 38 mm |
| Diameter (English) Well | 0.51 in. |
| Diameter (Metric) Well | 13 mm |