Learn More
Cytiva Strip Holder
Shop All Cytiva Products
Click to view available options
Size:
7cm
13cm
18cm
24cm
Quantity:
1 Each
6 Pack
Unit Size:
Each
Pack of 6
Specifications
Specifications
| For Use With (Equipment) | Ettan IPGphor 3 IEF System |
| Type | Strip Holder |
| Includes | 6 x Strip holder cover, 1 x 60mL strip holder cleaning solution, instructions |
| Quantity | 6 Pack |
| Size | 18cm |