Learn More
Cytiva Strip Holder
Product Code. 10167484 Shop All Cytiva Products
Click to view available options
Size:
7cm
13cm
18cm
24cm
Quantity:
1 Each
6 Pack
Unit Size:
Each
Pack of 6
Product Code. 10167484
Brand: Cytiva 80641744
Only null left
Specifications
For Use With (Equipment) Ettan IPGphor 3 IEF System
Type Strip Holder
Includes 6 x Strip holder cover, 1 x 60mL strip holder cleaning solution, instructions
Quantity 1 Each
Size 18cm