Learn More
Ansell™ String Knits™ 76-202 Series White Nylon Light Weight Gloves
Shop All Ansell Products
Click to view available options
Size:
6
7
8
9
10
11
Unit Size:
12 pairs
Description
- For general handling and delicate manipulation
- Can also be worn as liners
- Without seams to rub or irritate
- Ambidextrous
- Washable at 40°C
- Does not shrink, stiffen or lose shape
- Nylon liner
- Knitwrist cuff
Specifications
Specifications
| Back Material | Nylon |
| Construction | Seamless Knit |
| Cuff Material | Nylon |
| Product Type | Lightweight Gloves |
| Size | 8 |
| Style | 76-202 |
| Glove Cuff Color | White |
| Glove Cuff Style | Knit |
| Lining | Nylon |
| Quantity | 12 Pairs |
| Show More |