Learn More
Ansell™ Stringknits™ 76-200 Series Lightweight Nylon Gloves
Product Code. 12696971 Shop All Ansell Products
Click to view available options
Size:
Small
Medium
Large
X-Large
Unit Size:
12 pairs
Product Code. 12696971
Brand: Ansell™ 76200_7
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Permits longer, more comfortable wear with no weak points which might split or fray. Ansell Stringknits™ 76-200 Series Lightweight Nylon Gloves can be worn on their own or as liners.
  • Good value product protection
  • No seams to rub or irritate
  • Ambidextrous
  • Allows good air flow to the hand, preventing heat and sweat
  • Washable at 40°C without shrinkage or loss of shape
  • Soft, comfortable, flexible and low lint
  • Applications include general handling, delicate manipulation, fine assembly of dry parts
TRUSTED_SUSTAINABILITY
Specifications
Back Material Nylon
Construction Seamless Knit
Cuff Material Nylon
Coating Material Uncoated
Color White
Exterior Finish No Texture
Grade 13 gauge
Hand Ambidextrous
Material Nylon
Product Type Lightweight Gloves
Size Small
Glove Cuff Color White
Glove Cuff Style Knitwrist
Liner Material Nylon
Lining Lined
Quantity 12 Pairs
Resistant To Mechanical protection
Series 76-200