Learn More
MP Biomedicals™ Striker Bolt For FastPrep-24™
Product Code. 15658948 Shop All MP Biomedical Products
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Product Code. 15658948
Brand: MP Biomedicals™ 116005508
Striker Bolt is a part for the FastPrep-24™ 5G.
  • Samples and buffers are added to a 2.0 mL Lysing Matrix tube containing specialized Lysing Matrix beads.
  • The ergonomic design of the instrument ensures an easy loading of the sample tubes that remains securely sealed during the processing.
  • The homogenization speed and duration times are manually programmed or accessed from onboard memory.
  • After setting your speed and time with the touch of a button, just push run, and in less than a minute your samples are completely lysed.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) FastPrep-24 5G
Quantity 1 Ea.
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.