Learn More
Novus Biologicals™ Streptavidin Native Protein
Product Code. 18789473 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18789473 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18789473 Supplier Novus Biologicals™ Supplier No. NBP229370
Highly purified. Generating reliable and reproducible results.
Native Streptavidin. (Accession #: P22629) The Streptavidin Native Protein has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Formulation Lyophilized: 0.02 M Potassium Phosphate, 0.15M Sodium Chloride, pH 7.2 0.01% (w/v) Gentamicin Sulfate. 10mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free.
Quantity 1 mg
Content And Storage 4°C short term, −20°C long term. Avoid freeze-thaw cycles.
For Use With (Application) Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Purity or Quality Grade Chromatography
Product Type Streptavidin Protein