Learn More
Novus Biologicals™ Streptavidin Native Protein
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
Description
A full-length protein to Streptavidin. Source: Streptomyces avidinii The Streptavidin Native Protein has been validated for the following applications: Flow Cytometry.
Specifications
Specifications
| Formulation | Aqueous buffer, may contain carrier protein/stabilizer. |
| Quantity | 0.1 mg |
| Content And Storage | 4°C in the dark. |
| For Use With (Application) | FC |
| Product Type | Streptavidin Protein |