Learn More
Novus Biologicals™ Streptavidin Native Protein
Product Code. 18131315 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Product Code. 18131315
Brand: Novus Biologicals™ NBP229371
Highly purified. Generating reliable and reproducible results.
Native Streptavidin. (Accession #: P22629) The Streptavidin Native Protein has been validated for the following applications: Western Blot, ELISA, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Formulation 0.05 M Tris Chloride, 0.15M Sodium Chloride, 0.001M Magnesium Chloride, 0.0001M Zinc Chloride, 50% (v/v) Glycerol; pH 8.0, 10mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free; 0.05% Sodium Azide
Quantity 1 mg
Content And Storage 4°C. Do not freeze.
For Use With (Application) WB, ELISA, IHC
Product Type Streptavidin Protein