Learn More
BD Pharmingen™ FITC Streptavidin
Description
- This antibody is routinely tested by flow cytometric analysis.
- Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
- Host Species: Flow Cytometry
Specifications
Specifications
| Antigen | Streptavidin |
| Conjugate | FITC (Fluorescein) |
| Quantity | 0.5 mg |
| Regulatory Status | For research use only (RUO) |
| Storage Buffer | Aqueous buffered solution containing less than or equal to 0.09% sodium azide |
| Content And Storage | Store undiluted at 4°C. Protect from prolonged exposure to light. Do not freeze. |
| For Use With (Application) | Flow Cytometry |