Learn More
Equip Labo™ Straw Bench Splashes
Product Code. 12599225
Change view
Click to view available options
Depth (Metric):
750 mm
900 mm
Width (Metric):
120 cm
150 cm
180 cm
Top Style:
Glass Emalite
White Extruded Stoneware
White Melamine
Unit Size:
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Depth (Metric) Width (Metric) Top Style Unit Size
12599225 750 mm 150 cm Glass Emalite Each
12565326 750 mm 150 cm White Extruded Stoneware Each
12586506 750 mm 120 cm Glass Emalite Each
12587776 900 mm 150 cm Glass Emalite Each
12566486 900 mm 150 cm White Melamine Each
12565346 900 mm 180 cm White Melamine Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12599225 Supplier Equip Labo™ Supplier No. 8130
  • ONLINE EXCLUSIVE PRICE
    997.00€
    832.68€ / Pack of 1
    Save 164.32€
    16% Off
 Restricted Item
Estimated Shipment: 28-12-2026
Specifications
Product Type Straw Bench Splash
Depth (Metric) Shelf 150 mm
Depth (Metric) 750 mm
Dimensions (D x W x H) 750 x 1500 x 900 mm
Height (Metric) 900 mm
Load Bearing Capacity (Metric) 130 kg
Top Style Glass Emalite
Warranty 12 Months
Width (Metric) 150 cm