Learn More
Equip Labo™ Straw Bench Splashes
Brand: Equip Labo™ 8180
Additional Details : Weight : 0.80000kg
Specifications
Glass Emalite Worktop | |
Glass Emalite | |
Straw Bench Backsplash | |
900 mm | |
150 cm |
900 x 1500 x 900 mm | |
12 Months | |
900 mm | |
130 kg | |
150 mm |