Learn More
Equip Labo™ Straw Bench Splashes
Brand: Equip Labo™ 8197
Additional Details : Weight : 0.80000kg
Specifications
900 x 1800 x 900 mm | |
12 Months | |
900 mm | |
130 kg | |
150 mm |
White Melamine | |
Straw Bench Backsplash | |
900 mm | |
180 cm |