Learn More
Equip Labo™ Straw Bench Splashes
Product Code. 12565326
Click to view available options
Depth (Metric):
750 mm
900 mm
Width (Metric):
120 cm
150 cm
180 cm
Top Style:
Glass Emalite
White Extruded Stoneware
White Melamine
Unit Size:
Each
Product Code. 12565326
Brand: Equip Labo™ 8137
Specifications
Product Type Straw Bench Backsplash
Depth (Metric) Shelf 150 mm
Depth (Metric) 750 mm
Dimensions (D x W x H) 750 x 1500 x 900 mm
Height (Metric) 900 mm
Includes White Extruded Stoneware Worktop
Load Bearing Capacity (Metric) 130 kg
Top Style White Extruded Stoneware
Warranty 12 Months
Width (Metric) 150 cm