Learn More
Equip Labo™ Straw Bench Backsplashes
Brand:  Equip Labo™ 8304
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 12618832
  • 1310.00€ / Each
Estimated Shipment: 01-07-2024
Specifications
750 x 1800 x 900 mm
12 Months
900 mm
130 kg
150 mm
White Extruded Stoneware
Straw Bench Backsplash
750 mm
180 cm