Learn More
Equip Labo™ Straw Bench Backsplashes
Click to view available options
Dimensions (D x W x H):
750 x 900 x 900 mm
750 x 1200 x 900 mm
750 x 1800 x 900 mm
Top Style:
Glass Emalite
White Extruded Stoneware
White Melamine
Unit Size:
Each
Specifications
Specifications
| Product Type | Straw Bench Backsplash |
| Depth (Metric) Shelf | 150 mm |
| Depth (Metric) | 750 mm |
| Dimensions (D x W x H) | 750 x 1200 x 900 mm |
| Height (Metric) | 900 mm |
| Includes | White Extruded Stoneware Worktop |
| Load Bearing Capacity (Metric) | 130 kg |
| Top Style | White Extruded Stoneware |
| Warranty | 12 Months |
| Width (Metric) | 120 cm |