Learn More
Equip Labo™ Straw Bench Backsplashes
Brand:  Equip Labo™ 8146
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 12589235
  • 1135.00€ / Each
Estimated Shipment: 08-07-2024
Specifications
750 x 1800 x 900 mm
12 Months
900 mm
130 kg
150 mm
Glass Emalite
Straw Bench Backsplash
750 mm
180 cm