Learn More
Equip Labo™ Straw Bench Backsplashes
Brand: Equip Labo™ 8163
Additional Details : Weight : 0.80000kg
Specifications
750 x 900 x 900 mm | |
12 Months | |
900 mm | |
130 kg | |
150 mm |
White Melamine | |
Straw Bench Backsplash | |
750 mm | |
900 mm |