Learn More
Equip Labo™ Straw Bench Backsplashes
Product Code. 12547856
Change view
Click to view available options
Dimensions (D x W x H):
750 x 900 x 900 mm
750 x 1200 x 900 mm
750 x 1800 x 900 mm
Top Style:
Glass Emalite
White Extruded Stoneware
White Melamine
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (D x W x H) Top Style unitSize
12547856 750 x 900 x 900 mm Glass Emalite Each
12638832 750 x 900 x 900 mm White Extruded Stoneware Each
12589026 750 x 1200 x 900 mm White Melamine Each
12541766 750 x 1200 x 900 mm Glass Emalite Each
12597716 750 x 1200 x 900 mm White Extruded Stoneware Each
12589235 750 x 1800 x 900 mm Glass Emalite Each
12556486 750 x 1800 x 900 mm White Melamine Each
12575486 750 x 900 x 900 mm White Melamine Each
12618832 750 x 1800 x 900 mm White Extruded Stoneware Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 12547856 Supplier Equip Labo™ Supplier No. 8110
Specifications
Product Type Straw Bench Backsplash
Depth (Metric) Shelf 150 mm
Depth (Metric) 750 mm
Dimensions (D x W x H) 750 x 900 x 900 mm
Height (Metric) 900 mm
Load Bearing Capacity (Metric) 130 kg
Top Style Glass Emalite
Warranty 12 Months
Width (Metric) 900 mm