Learn More
Equip Labo™ Straw Bench Backsplashes
Brand:  Equip Labo™ 8120
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 12541766
  • 855.00€ / Each
Estimated Shipment: 05-07-2024
Specifications
750 x 1200 x 900 mm
12 Months
900 mm
130 kg
150 mm
Glass Emalite
Straw Bench Backsplash
750 mm
120 cm