Learn More
Karl Hecht™ Straight End Dropping Pipettes
Product Code. 30130490
Change view
Click to view available options
Length of approx. 80mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
30130490 Length of approx. 80mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30130490 Supplier Karl Hecht™ Supplier No. 42620010
  • ONLINE EXCLUSIVE PRICE
    7.55€
    3.23€ / Pack of 10
    Save 4.32€
    57% Off
View all promotions
Estimated Shipment: 20-11-2026
For accurate and precisely calibrated dropping in the laboratory.
  • Manufactured from clear glass
  • Solid finish
  • Without rubber teat
  • Length of approx. 80mm
  • Provided 50/pack
Specifications
Diameter (Metric) Outer Approx. 7-8 mm
Material Glass
For Use With (Application) Accurate dropping
Includes Straight end
Quantity 50/Pk.
Length (Metric) 80 mm
Type Dropping Pipette