Learn More
Gosselin™ Straight Assembled Sterile Container with Red screw Cap
Product Code. 15428794
Change view
Click to view available options
Capacity (Metric):
40 mL
60 mL
180 mL
Unit Size:
Pack of 100
Pack of 150
Pack of 50
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15428794 180 mL Pack of 50
15408794 40 mL Pack of 150
15418794 60 mL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15428794 Supplier Gosselin™ Supplier No. BTP53C002_CE_IVD
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Polypropylene assembled straight container with screw cap
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Sterility Sterile
Capacity (Metric) 180 mL
Closure Color Red
Closure Type Screw Cap with Inner Lip
Color Clear