Learn More
Thermo Scientific™ Storage Vials
Product Code. 12468676
Click to view available options
Color:
Amber
Clear
White
White, Clear
Quantity:
100
100 Caps
100 Septa
100 Vials
200
200 (vial and caps are sold separately)
200 Vials
Unit Size:
Pack of 100
Pack of 200
Volume (Metric):
2 mL
4 mL
8 mL
12 mL
16 mL
22 mL
40 mL
Closure Size:
13-425
15 mm
15-425
18 mm
18-400
20-400
24 mm
24-400
8-425
Product Code. 12468676
Brand: Thermo Scientific™ B799526
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Store your samples in these vials to eliminate leaching of ions while maintaining a consistent pH for the life of your sample. Thermo Scientific™ National™ and Thermo Scientific™ Chromacol™ Sample Storage Screw Thread Vials are manufactured from superior quality 33 expansion borosilicate clear (Type 1, Class A) or 51A amber (Type 1, Class B) glass. Storage vials range from 2mL to 40mL.
  • PTFE-lined solid-top storage caps
  • PTFE on film/foam backing offers broad chemical resistance
  • Not autoclavable
Order as individual sample storage vials and closures or as unassembled convenience kits.
Unassembled convenience kits saves time during sample preparation.
  • Solid-top convenience kits include matched quantities of vials and screw caps with pre-assembled septa.
  • Open-top convenience kits contain shrink-wrapped vials and separately packaged caps and septa in polybags.
TRUSTED_SUSTAINABILITY
Specifications
Description Septa for 24-400 Screw Cap
Color White, Clear
Material PTFE, Silicone
Quantity 100
Thickness (Metric) 3.3 mm
Type Septum Only