Learn More
BOLA Stoppers
Product Code. 10013231
Change view
Click to view available options
Size:
19 mm
26 mm
35 mm
52 mm
NS 14/23
NS 34/35
NS 45/40
Unit Size:
Each
Pack of 3
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
10013231 19 mm Each
11753269 NS 14/23 Pack of 3
10186110 NS 34/35 Each
10399231 NS 45/40 Each
10776172 26 mm Each
10795982 35 mm Each
10699211 52 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 10013231 Supplier BOLA Supplier No. H93704
  • ONLINE EXCLUSIVE PRICE
    26.25€
    21.40€ / Pack of 1
    Save 4.85€
    18% Off
Estimated Shipment: 03-11-2026
Stoppers PTFE, with grip
  • Made of PTFE, with ground joint and sealing rings on the outside knurled or hexagonal grip.
  • Compared with glass stoppers, they are easily removable and can be used without grease.
  • The stoppers can expand under heat which might lead to a breaking of the ground joint sockets.
Specifications
Size 19 mm
Autoclavable Autoclavable
Color White