Learn More
Tocris Bioscience™ STO-609 acetate
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
Description
STO-609 acetate is a selective, cell-permeable inhibitor of Ca2+-calmodulin-dependent protein kinase kinase (Ki values are 80 and 15 ng/ml for inhibition of CaM-KKα and CaM-KKβ respectively); competes for the ATP-binding site. Displays > 80-fold selectivity over CaMK1, CaMK2, CaMK4, MLCK, PKC, PKA and p42 MAPK.
Specifications
Specifications
| Chemical Name or Material | 7-Oxo-7H-benzimidazo[2,1-a]benz[de]isoquinoline-3-carboxylic acid acetate |
| CAS | 1173022-21-3 |
| Assay Percent Range | >98% |
| Quantity | 1 mg |
| Solubility | Soluble to 10mM with sonication in DMSO |
| Molecular Formula | C19H10N2O3.C2H4O2 |
| Formula Weight | Observed MW: 374.35 |